Product Details

SNP ID
rs2287499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7688850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTCAGCTGTGTCCCAGGAGCTA[C/G]GGGAGGGGGACCCAGTTTCTCTCTC
Phenotype
MIM: 191170 MIM: 612661
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
There are no transcripts associated with this gene.

Gene
WRAP53
Gene Name
WD repeat containing antisense to TP53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143990.1 357 Missense Mutation CGG,GGG R68G NP_001137462.1
NM_001143991.1 357 Missense Mutation CGG,GGG R68G NP_001137463.1
NM_001143992.1 357 Missense Mutation CGG,GGG R68G NP_001137464.1
NM_018081.2 357 Missense Mutation CGG,GGG R68G NP_060551.2
XM_011523952.2 357 UTR 5 XP_011522254.1
XM_017024812.1 357 Missense Mutation CGG,GGG R68G XP_016880301.1

View Full Product Details