Product Details

SNP ID
rs2275074
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:156068301 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TCGTGGTGCAGTGGGGGCCCTCCTG[A/G]TGTTTGACCTAACCAAGCACCAGAC
Phenotype
MIM: 610389 MIM: 611007 MIM: 612942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR2 PubMed Links

Gene Details

Gene
LAMTOR2
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
There are no transcripts associated with this gene.

Gene
MEX3A
Gene Name
mex-3 RNA binding family member A
There are no transcripts associated with this gene.

Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020387.3 523 Missense Mutation ATG,GTG M91V NP_065120.2

View Full Product Details