Product Details

SNP ID
rs2296767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70042573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTGGCTGCCTGTGGGAGCAAAG[C/T]AGCAGTAAGCTAAAGAAGGAGGCAA
Phenotype
MIM: 300925 MIM: 300451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AWAT2 PubMed Links

Gene Details

Gene
AWAT2
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002254.1 Intron NP_001002254.1
XM_011530876.2 Intron XP_011529178.1
Gene
EDA
Gene Name
ectodysplasin A
There are no transcripts associated with this gene.

View Full Product Details