Product Details

SNP ID
rs2293969
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128818719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGGCCGGCCTGACCCTGGCGT[C/T]GGGCGCGGGGCTGGGTGCGGTCGTC
Phenotype
MIM: 600440 MIM: 616218
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ENDOG PubMed Links

Gene Details

Gene
ENDOG
Gene Name
endonuclease G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004435.2 318 Missense Mutation TCG,TTG S12L NP_004426.2
XM_011518347.2 318 Missense Mutation TCG,TTG S101L XP_011516649.1
Gene
SPOUT1
Gene Name
SPOUT domain containing methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016390.3 318 Intron NP_057474.2
XM_017014804.1 318 Intron XP_016870293.1
Gene
TBC1D13
Gene Name
TBC1 domain family member 13
There are no transcripts associated with this gene.

View Full Product Details