Product Details

SNP ID
rs2294017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49237280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTGCATGGTGAGTGGCCCTCCTC[T/C]TAATGCACACATCCTCTTTCTTCCT
Phenotype
MIM: 300110 MIM: 300859
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CACNA1F PubMed Links

Gene Details

Gene
CACNA1F
Gene Name
calcium voltage-gated channel subunit alpha1 F
There are no transcripts associated with this gene.

Gene
CCDC22
Gene Name
coiled-coil domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014008.4 Intron NP_054727.1
XM_005272599.3 Intron XP_005272656.1

View Full Product Details