Product Details

SNP ID
rs2297792
Assay Type
Validated
NCBI dbSNP Submissions
85
Location
Chr.1:156041653 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CTGCTGAGGGTGCTGGGGTCCGGGA[C/T]ATCCCAAAGGAGCCAAGGCTGTAGG
Phenotype
MIM: 605440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBQLN4 PubMed Links

Gene Details

Gene
UBQLN4
Gene Name
ubiquilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304342.1 Intron NP_001291271.1
NM_020131.4 Intron NP_064516.2
XM_005245348.3 Intron XP_005245405.1

View Full Product Details