Product Details

SNP ID
rs2295279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107704212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACACTCTCCCATTTCAAAGCTTC[A/G]TTGAGACATTTTCTTTTCCTTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCML4 PubMed Links

Gene Details

Gene
SCML4
Gene Name
sex comb on midleg-like 4 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286408.1 2226 UTR 3 NP_001273337.1
NM_001286409.1 2226 UTR 3 NP_001273338.1
NM_198081.4 2226 UTR 3 NP_932347.2
XM_011535703.2 2226 UTR 3 XP_011534005.1
XM_011535704.2 2226 UTR 3 XP_011534006.1
XM_011535706.1 2226 UTR 3 XP_011534008.1
XM_011535707.2 2226 Intron XP_011534009.1
XM_011535709.2 2226 UTR 3 XP_011534011.1
XM_011535710.2 2226 UTR 3 XP_011534012.1
XM_017010677.1 2226 UTR 3 XP_016866166.1
XM_017010678.1 2226 UTR 3 XP_016866167.1
XM_017010679.1 2226 UTR 3 XP_016866168.1
XM_017010680.1 2226 UTR 3 XP_016866169.1
XM_017010681.1 2226 UTR 3 XP_016866170.1
XM_017010682.1 2226 UTR 3 XP_016866171.1
XM_017010683.1 2226 UTR 3 XP_016866172.1
XM_017010684.1 2226 Intron XP_016866173.1
XM_017010685.1 2226 UTR 3 XP_016866174.1
XM_017010686.1 2226 UTR 3 XP_016866175.1
XM_017010687.1 2226 UTR 3 XP_016866176.1
XM_017010688.1 2226 Intron XP_016866177.1

View Full Product Details