Product Details

SNP ID
rs2295480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:733707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCCAGAGGCAGGTCAGGAAATG[G/T]GCACCTATCTGCAGTTCCAGTGAAA
Phenotype
MIM: 611627
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM57A PubMed Links

Gene Details

Gene
FAM57A
Gene Name
family with sequence similarity 57 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318006.1 Intron NP_001304935.1
NM_001318007.1 Intron NP_001304936.1
NM_001318008.1 Intron NP_001304937.1
NM_024792.2 Intron NP_079068.1

View Full Product Details