Product Details

SNP ID
rs2293020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43197439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAACCCAGCCCAGCGGCGCCA[A/G]CCAGAACGTGTCATGATGTCCCTTC
Phenotype
MIM: 614531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASGEF1A PubMed Links

Gene Details

Gene
RASGEF1A
Gene Name
RasGEF domain family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282862.1 Intron NP_001269791.1
NM_145313.3 Intron NP_660356.2
XM_005271809.3 Intron XP_005271866.1
XM_011539500.2 Intron XP_011537802.1

View Full Product Details