Product Details

SNP ID
rs2307314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127604774 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCTGGGCCGCATGCGCCGTGGG[C/T]TCCTGTATGGTAGGTCCAGTTGTCT
Phenotype
MIM: 116945 MIM: 608336
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM2 PubMed Links

Gene Details

Gene
MCM2
Gene Name
minichromosome maintenance complex component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004526.3 513 Missense Mutation CTC,TTC L135F NP_004517.2
Gene
TPRA1
Gene Name
transmembrane protein adipocyte associated 1
There are no transcripts associated with this gene.

View Full Product Details