Product Details

SNP ID
rs2334102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50531323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGGCGGATCAAGGGTGTGGGGG[C/G]CTAGAACTGCGTGGGAGCCTGGTGG
Phenotype
MIM: 611230 MIM: 604272 MIM: 131222
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NCAPH2 PubMed Links

Gene Details

Gene
NCAPH2
Gene Name
non-SMC condensin II complex subunit H2
There are no transcripts associated with this gene.

Gene
ODF3B
Gene Name
outer dense fiber of sperm tails 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014440.3 Intron NP_001014440.2
Gene
SCO2
Gene Name
SCO2 cytochrome c oxidase assembly protein
There are no transcripts associated with this gene.

Gene
TYMP
Gene Name
thymidine phosphorylase
There are no transcripts associated with this gene.

View Full Product Details