Product Details
- SNP ID
-
rs2328237
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:18234632 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTCAGCCAAAAAGCCATTATTA[C/T]GAATTCCTTCAATTTTCTGACAGCA
- Phenotype
-
MIM: 125264
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEK
PubMed Links
Gene Details
- Gene
- DEK
- Gene Name
- DEK proto-oncogene
View Full Product Details