Product Details
- SNP ID
-
rs3026122
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:5385446 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCATGTGGCTTTTAATTGACAGTC[A/G]CTCAGCCATTTCTAAGCAGATATAG
- Phenotype
-
MIM: 602552
MIM: 603616
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NUP88
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76828367] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUP88
- Gene Name
- nucleoporin 88
- Gene
- RABEP1
- Gene Name
- rabaptin, RAB GTPase binding effector protein 1
View Full Product Details