Product Details

SNP ID
rs2343694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30833635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGTCCTTTAGTTCCATTTTCTG[A/G]TGTTGAATACAAGCACCACGAACAA
Phenotype
MIM: 605795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX15 PubMed Links

Gene Details

Gene
TEX15
Gene Name
testis expressed 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031271.3 Intron NP_112561.2
XM_005273575.3 Intron XP_005273632.1
XM_005273576.3 Intron XP_005273633.1
XM_006716369.3 Intron XP_006716432.1
XM_011544588.2 Intron XP_011542890.1
XM_011544589.2 Intron XP_011542891.1
XM_011544590.2 Intron XP_011542892.1
XM_011544591.2 Intron XP_011542893.1
XM_011544592.2 Intron XP_011542894.1
XM_011544593.2 Intron XP_011542895.1
XM_011544594.2 Intron XP_011542896.1

View Full Product Details