Product Details

SNP ID
rs2404814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4380458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAGCTTCTCTGCTTACACACC[A/C]ACCCAGCTGCCTGCCTGCCCCCTCT
Phenotype
MIM: 611668 MIM: 608843
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CORO7 PubMed Links

Gene Details

Gene
CORO7
Gene Name
coronin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201472.1 Intron NP_001188401.1
NM_001201473.1 Intron NP_001188402.1
NM_024535.4 Intron NP_078811.3
Gene
CORO7-PAM16
Gene Name
CORO7-PAM16 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201479.1 Intron NP_001188408.1
Gene
VASN
Gene Name
vasorin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138440.2 Intron NP_612449.2

View Full Product Details