Product Details
- SNP ID
-
rs2427249
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62148126 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACCCGAGAAGGGGAGCATGGGG[A/G]CCTGTCTTTACATCCGCAGGGATAG
- Phenotype
-
MIM: 606607
MIM: 606472
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PSMA7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114427846] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PSMA7
- Gene Name
- proteasome subunit alpha 7
There are no transcripts associated with this gene.
- Gene
- SS18L1
- Gene Name
- SS18L1, nBAF chromatin remodeling complex subunit
View Full Product Details