Product Details

SNP ID
rs2437329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40756224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGCCTGCTGAGCAGGAAGAGGG[C/T]GAGAGATACCCCAGTTCCTTCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN7 PubMed Links

Gene Details

Gene
NSUN7
Gene Name
NOP2/Sun RNA methyltransferase family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017008611.1 Intron XP_016864100.1
XM_017008612.1 Intron XP_016864101.1
XM_017008613.1 Intron XP_016864102.1
XM_017008614.1 Intron XP_016864103.1
XM_017008615.1 Intron XP_016864104.1

View Full Product Details