Product Details
- SNP ID
-
rs2257914
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30152786 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACAAGAAAATAAAACATGGTCACT[A/C]TTCCATCCCATTTCTCAGATGTCAC
- Phenotype
-
MIM: 605701
MIM: 616976
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17194411] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRIM10
- Gene Name
- tripartite motif containing 10
- Gene
- TRIM40
- Gene Name
- tripartite motif containing 40
There are no transcripts associated with this gene.
View Full Product Details