Product Details

SNP ID
rs2532662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117488179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGCAGCATGGCTCAACATGCG[A/G]TAGGCGGAAGATTGATGCAGGAGTA
Phenotype
MIM: 607637
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMX2OS PubMed Links

Gene Details

Gene
EMX2OS
Gene Name
EMX2 opposite strand/antisense RNA
There are no transcripts associated with this gene.

View Full Product Details