Product Details

SNP ID
rs2503699
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:2187100 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCACAGTGGGCCTGCGTGGGCG[T/C]GGTGCGGGGCAACTGCCCATGGAGC
Phenotype
MIM: 615183 MIM: 176982
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FAAP20 PubMed Links

Gene Details

Gene
FAAP20
Gene Name
Fanconi anemia core complex associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146310.1 251 Intron NP_001139782.1
NM_001256945.1 251 Intron NP_001243874.1
NM_001256946.1 251 Intron NP_001243875.1
NM_001256947.1 251 Intron NP_001243876.1
NM_001282670.1 251 Intron NP_001269599.1
NM_001282671.1 251 UTR 5 NP_001269600.1
NM_001282672.1 251 UTR 5 NP_001269601.1
NM_001282673.1 251 Intron NP_001269602.1
NM_182533.2 251 Intron NP_872339.2
XM_006710419.3 251 Intron XP_006710482.1
XM_006710421.3 251 Intron XP_006710484.1
XM_011540914.2 251 Intron XP_011539216.1
XM_011540921.2 251 Intron XP_011539223.1
XM_011540922.1 251 Intron XP_011539224.1
XM_017000553.1 251 Intron XP_016856042.1
XM_017000554.1 251 Intron XP_016856043.1
XM_017000555.1 251 UTR 5 XP_016856044.1
XM_017000556.1 251 UTR 5 XP_016856045.1
XM_017000557.1 251 Intron XP_016856046.1
Gene
LOC100506504
Gene Name
uncharacterized LOC100506504
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
There are no transcripts associated with this gene.

View Full Product Details