Product Details

SNP ID
rs2550051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31134559 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTCCCTCAAAACCACCTCCAGGA[C/T]GGGAACGGTGGCTCACACCTGTAAT
Phenotype
MIM: 609912 MIM: 610560 MIM: 600823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT8 PubMed Links

Gene Details

Gene
KAT8
Gene Name
lysine acetyltransferase 8
There are no transcripts associated with this gene.

Gene
PRSS36
Gene Name
protease, serine 36
There are no transcripts associated with this gene.

Gene
PRSS8
Gene Name
protease, serine 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002773.4 Intron NP_002764.1

View Full Product Details