Product Details

SNP ID
rs2576431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127850071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTCACAATCCTACCATCAGCG[G/T]ACAGAAGTTCCAGCTGTTTCATATT
Phenotype
MIM: 606017
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC107985803 PubMed Links

Gene Details

Gene
LOC107985803
Gene Name
uncharacterized LOC107985803
There are no transcripts associated with this gene.

Gene
POLR2D
Gene Name
RNA polymerase II subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004805.3 Intron NP_004796.1

View Full Product Details