Product Details

SNP ID
rs2622924
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:47352791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGCCCATGTAGTCTGCTGTAC[A/G]TTATAGAAATGGGTACAGTTATTTA
Phenotype
MIM: 191710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMPK1 PubMed Links

Gene Details

Gene
CMPK1
Gene Name
cytidine/uridine monophosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136140.1 Intron NP_001129612.1
NM_016308.2 Intron NP_057392.1

View Full Product Details