Product Details

SNP ID
rs2603141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108549439 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCAAAGCTGCAGTACGGACTTTC[C/T]TCTTTAGGGAGAGAAGATTAGAAAT
Phenotype
MIM: 610643 MIM: 609929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1524 PubMed Links

Gene Details

Gene
KIAA1524
Gene Name
KIAA1524
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020890.2 24 Intron NP_065941.2
XM_006713716.3 24 Intron XP_006713779.1
XM_006713717.2 24 UTR 3 XP_006713780.1
XM_011513056.1 24 UTR 3 XP_011511358.1
XM_011513057.2 24 UTR 3 XP_011511359.1
Gene
MYH15
Gene Name
myosin heavy chain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014981.1 24 Intron NP_055796.1
XM_011512559.2 24 UTR 5 XP_011510861.1
XM_017005922.1 24 Intron XP_016861411.1

View Full Product Details