Product Details

SNP ID
rs2562396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:90805143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTTGATTCTTTTCTTTCTGGGA[A/C]ACCACGTATTGATTTCTGGCAGCTG
Phenotype
MIM: 600085
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYK PubMed Links

Gene Details

Gene
SYK
Gene Name
spleen associated tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135052.3 Intron NP_001128524.1
NM_001174167.2 Intron NP_001167638.1
NM_001174168.2 Intron NP_001167639.1
NM_003177.6 Intron NP_003168.2
XM_005252147.3 Intron XP_005252204.1
XM_011518946.2 Intron XP_011517248.1

View Full Product Details