Product Details
- SNP ID
-
rs2637666
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116497047 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCACCTTTTAGTCTGTCTTGCAA[A/G]AATGTAACATCAGCCGCCAAATGAA
- Phenotype
-
MIM: 614137
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRAPPC3L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114678052] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRAPPC3L
- Gene Name
- trafficking protein particle complex 3 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001139444.2 |
549 |
Silent Mutation |
TTC,TTT |
F151F |
NP_001132916.1 |
View Full Product Details