Product Details

SNP ID
rs2706346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132569065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAATTCACAATCAAAAGTTAACA[A/G]GCATCCAAAGAAGCAGGGAAATAGG
Phenotype
MIM: 604040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD50 PubMed Links

Gene Details

Gene
RAD50
Gene Name
RAD50 double strand break repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005732.3 Intron NP_005723.2

View Full Product Details