Product Details

SNP ID
rs2742215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78951692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTTTTAGTCATAACTTTGTGC[A/G]TGCCATTTTTATTATCAGGAACATC
Phenotype
MIM: 300529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
P2RY10 PubMed Links

Gene Details

Gene
P2RY10
Gene Name
purinergic receptor P2Y10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324218.1 Intron NP_001311147.1
NM_001324221.1 Intron NP_001311150.1
NM_001324225.1 Intron NP_001311154.1
NM_014499.3 Intron NP_055314.1
NM_198333.2 Intron NP_938147.1

View Full Product Details