Product Details

SNP ID
rs2736172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31623121 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGTGGTTCCCTGTCTTTGGACA[C/T]GTAAGAATTGGAGGAAAATAAATGT
Phenotype
MIM: 601833 MIM: 142580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIF1 PubMed Links

Gene Details

Gene
AIF1
Gene Name
allograft inflammatory factor 1
There are no transcripts associated with this gene.

Gene
PRRC2A
Gene Name
proline rich coiled-coil 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004638.3 Intron NP_004629.3
NM_080686.2 Intron NP_542417.2
XM_017011274.1 Intron XP_016866763.1
Gene
SNORA38
Gene Name
small nucleolar RNA, H/ACA box 38
There are no transcripts associated with this gene.

View Full Product Details