Product Details
- SNP ID
-
rs2756105
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:99787285 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCCTTCCCCTTATCCTTAGCAAC[C/T]GCTAAATCTCTTTTCTGTCTTTATA
- Phenotype
-
MIM: 601107
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7393105] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCC2
- Gene Name
- ATP binding cassette subfamily C member 2
View Full Product Details