Product Details

SNP ID
rs3793900
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:86959076 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGCTACCTGTCTGTGCACGCACA[A/C]ACACATTCCCAAGCATACCAGCCTC
Phenotype
MIM: 608925 MIM: 602998
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADIRF PubMed Links

Gene Details

Gene
ADIRF
Gene Name
adipogenesis regulatory factor
There are no transcripts associated with this gene.

Gene
MMRN2
Gene Name
multimerin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024756.2 Intron NP_079032.2
XM_005270153.1 Intron XP_005270210.1
XM_006717970.3 Intron XP_006718033.1
Gene
SNCG
Gene Name
synuclein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003087.2 Intron NP_003078.2
XM_011540105.2 Intron XP_011538407.1

View Full Product Details