Product Details

SNP ID
rs3737155
Assay Type
Validated
NCBI dbSNP Submissions
61
Location
Chr.1:10399722 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GCTTTGTGGTAAGCGGCGTGGGCGC[A/G]TTGTCTTCTCTCTGGTTCCCGGGCG
Phenotype
MIM: 172200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGD PubMed Links

Gene Details

Gene
PGD
Gene Name
phosphogluconate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304451.1 Intron NP_001291380.1
NM_001304452.1 Intron NP_001291381.1
NM_002631.3 Intron NP_002622.2

View Full Product Details