Product Details

SNP ID
rs77794544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69341435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCACGTTTTGAATCCCAGAAAG[A/G]CCAGAACTGTATTTTTTCCGTCTTT
Phenotype
MIM: 606979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG8 PubMed Links

Gene Details

Gene
COG8
Gene Name
component of oligomeric golgi complex 8
There are no transcripts associated with this gene.

Gene
NIP7
Gene Name
NIP7, nucleolar pre-rRNA processing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199434.1 Intron NP_001186363.1
NM_016101.4 Intron NP_057185.1
Gene
TMED6
Gene Name
transmembrane p24 trafficking protein 6
There are no transcripts associated with this gene.

View Full Product Details