Product Details

SNP ID
rs61747114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72376855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGAGCAATTGAAAATCATCCC[A/G]AAAGATCTGCATTTTAGAAATACCA
Phenotype
MIM: 606869
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEXA PubMed Links

Gene Details

Gene
HEXA
Gene Name
hexosaminidase subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000520.5 Intron NP_000511.2
NM_001318825.1 Intron NP_001305754.1
Gene
HEXA-AS1
Gene Name
HEXA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details