Product Details

SNP ID
rs55879823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95585599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATTCAGGAAGCAGCACTGGTG[C/T]AGACCCACTTTGATCCCGTAAAGTG
Phenotype
MIM: 602527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDK4 PubMed Links

Gene Details

Gene
PDK4
Gene Name
pyruvate dehydrogenase kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002612.3 1598 UTR 3 NP_002603.1

View Full Product Details