Product Details

SNP ID
hCV22274564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111866378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTCTTCCCCATTCATGCTATC[C/T]TAGTAAAGTAGTCCTAGATCTGTCA
Phenotype
MIM: 606723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPKAPK5 PubMed Links

Gene Details

Gene
MAPKAPK5
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003668.3 Intron NP_003659.2
NM_139078.2 Intron NP_620777.1

View Full Product Details