Product Details

SNP ID
rs2273077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62797470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCAGAGACCCCTGCAGTGAGGC[C/T]CGTCCTTAAGGAGGACATCCCGGGG
Phenotype
MIM: 611157 MIM: 606459
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGBP PubMed Links

Gene Details

Gene
MRGBP
Gene Name
MRG/MORF4L binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018270.5 Intron NP_060740.1
Gene
OGFR
Gene Name
opioid growth factor receptor
There are no transcripts associated with this gene.

Gene
OGFR-AS1
Gene Name
OGFR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details