Product Details

SNP ID
rs7278223
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25881907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAACACCCATAATTTTCTCCCC[C/T]ACTTTGACATCTTGGAGCAGAACGC
Phenotype
MIM: 104760
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
APP PubMed Links

Gene Details

Gene
APP
Gene Name
amyloid beta precursor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000484.3 Intron NP_000475.1
NM_001136016.3 Intron NP_001129488.1
NM_001136129.2 Intron NP_001129601.1
NM_001136130.2 Intron NP_001129602.1
NM_001136131.2 Intron NP_001129603.1
NM_001204301.1 Intron NP_001191230.1
NM_001204302.1 Intron NP_001191231.1
NM_001204303.1 Intron NP_001191232.1
NM_201413.2 Intron NP_958816.1
NM_201414.2 Intron NP_958817.1

View Full Product Details