Product Details

SNP ID
rs79942400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4247273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCTTCTTTGGAACCCTTCTTTC[C/T]CAGCGGCCTTCCGCGAGATGGGGCT
Phenotype
MIM: 605816
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC94 PubMed Links

Gene Details

Gene
CCDC94
Gene Name
coiled-coil domain containing 94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018074.4 Intron NP_060544.2
Gene
EBI3
Gene Name
Epstein-Barr virus induced 3
There are no transcripts associated with this gene.

View Full Product Details