Product Details

SNP ID
rs55711976
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41797048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAATATAACCAGGGCACTAAA[A/C]CCTGTCTTTGACCACCAACCACAAG
Phenotype
MIM: 609142
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CEACAM3 PubMed Links

Gene Details

Gene
CEACAM3
Gene Name
carcinoembryonic antigen related cell adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277163.2 Intron NP_001264092.1
NM_001815.4 Intron NP_001806.2

View Full Product Details