Product Details
- SNP ID
-
rs75924208
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:40301754 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATGGAGGAGACCGCAGTGTCCAC[A/G]CGTCATTAATTCCCAAGCTGAGGAA
- Phenotype
-
MIM: 120260
MIM: 606480
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COL9A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150450056] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL9A2
- Gene Name
- collagen type IX alpha 2
- Gene
- ZMPSTE24
- Gene Name
- zinc metallopeptidase STE24
There are no transcripts associated with this gene.
View Full Product Details