Product Details
- SNP ID
-
rs7146118
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:92926489 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTCCCTGAGCCAGGGATCATGGA[A/C]TCCCAAAGACAAAAGCAAATACCCT
- Phenotype
-
MIM: 118910
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CHGA
PubMed Links
Gene Details
- Gene
- CHGA
- Gene Name
- chromogranin A
View Full Product Details