Product Details

SNP ID
rs61732498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3089309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCGGGCTCCGCATGGGTAGCCA[C/G]GTGCCGCCGCAGATGCGCGTTGCGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 2052 Missense Mutation CTG,GTG L315V NP_001269344.1
NM_001282416.1 2052 Missense Mutation CTG,GTG L572V NP_001269345.1
NM_032805.2 2052 Missense Mutation CTG,GTG L709V NP_116194.2
XM_017023791.1 2052 Missense Mutation CTG,GTG L709V XP_016879280.1
XM_017023792.1 2052 Missense Mutation CTG,GTG L677V XP_016879281.1

View Full Product Details