Product Details

SNP ID
rs36049318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136496450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCTCCACTCAGGAAGCTCCGG[C/G]CCAGGTGGCCGCTGGCTGCTGAGCT
Phenotype
MIM: 190198
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOTCH1 PubMed Links

Gene Details

Gene
NOTCH1
Gene Name
notch 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017617.4 6581 Missense Mutation GCC,GGC A2430G NP_060087.3
XM_011518717.2 6581 Missense Mutation GCC,GGC A2189G XP_011517019.2

View Full Product Details