Product Details

SNP ID
rs28578906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57238122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAAAAGTTATAGATGAGGTAAA[C/T]GTTTATTTTATTTTGCTTGATTAAA
Phenotype
MIM: 604299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012096.2 Intron NP_036228.1
XM_011533583.2 Intron XP_011531885.1

View Full Product Details