Product Details

SNP ID
rs9333521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43587240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATCTCAAGGTTGCCTCTGTTTA[C/T]TGCCTGCATGAATGATCCTTATACT
Phenotype
MIM: 603968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLH PubMed Links

Gene Details

Gene
POLH
Gene Name
DNA polymerase eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291969.1 Intron NP_001278898.1
NM_001291970.1 Intron NP_001278899.1
NM_006502.2 Intron NP_006493.1
XM_005249186.3 Intron XP_005249243.1
XM_011514698.2 Intron XP_011513000.1

View Full Product Details