Product Details

SNP ID
rs34890926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106924644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTCGATCTCTGTGCCGAGTAC[C/T]ATCCAGAGCCGGGATGTGAGGGGTT
Phenotype
MIM: 605415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKK2 PubMed Links

Gene Details

Gene
DKK2
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014421.2 1135 Missense Mutation AGT,GGT S144G NP_055236.1

View Full Product Details