Product Details

SNP ID
rs6739001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174748749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTCTTCTGTAAAAATCTTTTTG[A/T]CTTGCTTTTCTCTGGATGGTCTTTT
Phenotype
MIM: 100690
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHRNA1 PubMed Links

Gene Details

Gene
CHRNA1
Gene Name
cholinergic receptor nicotinic alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000079.3 1151 Missense Mutation GAC,GTC D358V NP_000070.1
NM_001039523.2 1151 Missense Mutation GAC,GTC D383V NP_001034612.1
XM_017003256.1 1151 Missense Mutation GAC,GTC D390V XP_016858745.1
XM_017003257.1 1151 Missense Mutation GAC,GTC D365V XP_016858746.1

View Full Product Details