Product Details

SNP ID
rs35375661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58477366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCCCTGGCCCGGCTGCGTGAG[C/T]TGTGTTGCCAGTGGCTGCAGCCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF324 PubMed Links

Gene Details

Gene
ZNF324
Gene Name
zinc finger protein 324
There are no transcripts associated with this gene.

Gene
ZNF446
Gene Name
zinc finger protein 446
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304453.1 529 Silent Mutation CTG,TTG L50L NP_001291382.1
NM_017908.3 529 Silent Mutation CTG,TTG L50L NP_060378.1
XM_005259052.4 529 Silent Mutation CTG,TTG L50L XP_005259109.1
XM_005259053.4 529 Silent Mutation CTG,TTG L50L XP_005259110.1
XM_006723266.3 529 Silent Mutation CTG,TTG L50L XP_006723329.1
XM_017026949.1 529 Silent Mutation CTG,TTG L50L XP_016882438.1
XM_017026950.1 529 Silent Mutation CTG,TTG L50L XP_016882439.1

View Full Product Details