Product Details

SNP ID
rs111308632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40669856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATAGAGGAGGGGATACAGGA[A/G]GGACATGCAGGGTGTCTGCCCAGCA
Phenotype
MIM: 604018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUMBL PubMed Links

Gene Details

Gene
NUMBL
Gene Name
NUMB like, endocytic adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289979.1 Intron NP_001276908.1
NM_001289980.1 Intron NP_001276909.1
NM_004756.4 Intron NP_004747.1

View Full Product Details